Merck
[Permeability of pannexin 1 channels to large anions].
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova (2013-03-07)
R A Romanov, O A Rogachevskaia, A S Kolesnikova, A A Khokhlov, S S Kolesnikov
ABSTRACT
It is widely accepted that ATP secretion in diverse cells involves pannexin 1 (Panx1) that functions as an ATP-permeable channel. We analyzed the permeability of Panx1 channels heterologically expressed in HEK-293 cells to a variety of anions, including ATP. As was demonstrated in electrophysiological experiments, relative permeabilities of studied species follow the sequence: C1- > MeSO4 > gluconate > HEPES. This sequence suggests that ATP, which is more than twice as large as HEPES by mass, is most likely to be negligibly permeable to Panx1 channels. This inference was verified in experiments, where ATP secretion from Panx1-positive HEK-293 cells was assayed with the ATP-biosensor approach. It was shown that the heterologous expression of Panx1 in HEK-293, which normally are not ATP-secretive, did not endow transfected cells with the ability to liberate ATP in response to stimulation. Our data indicate that Panx1 alone forms anion channels with too low ATP permeability to mediate the secretory function. Nevertheless, the possibility still remains that certain ATP-permeable channels are heteromers of Panx1 and some other channel subunit(s).
Sigma-Aldrich
D-Gluconic acid solution, 49-53 wt. % in H2O
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
HEPES sodium salt solution, BioReagent, 1M, suitable for cell culture
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
D-Gluconic acid sodium salt, ≥99% (HPLC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium gluconate, meets USP testing specifications
Sigma-Aldrich
HEPES sodium salt, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
SAFC
HEPES
Supelco
D-Gluconic acid sodium salt, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
HEPES sodium salt, Vetec, reagent grade, 96%
SAFC
HEPES sodium salt
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, 99.5-100.5% (calc. to the dried substance), meets analytical specification of Ph. Eur., BP,USP, FCC
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium D-gluconate hydrate, ≥98% (HPLC)
Sigma-Aldrich
Potassium gluconate, 97.0-103.0% dry basis, meets USP testing specifications
Sigma-Aldrich
Magnesium D-gluconate hydrate, 98.0-102% anhydrous basis, meets USP testing specifications