Merck
  • Quantification of arsenic in dialysate solution and scalp hair samples of kidney failure patients by cloud point extraction and electrothermal atomic absorption spectroscopy.
Quantification of arsenic in dialysate solution and scalp hair samples of kidney failure patients by cloud point extraction and electrothermal atomic absorption spectroscopy.
Journal of AOAC International (2013-03-05)
Jameel A Baig, Tasneem G Kazi, Mohammad Balal Arain, Hassan Imran Afridi, Khalida Perveen Mahar
ABSTRACT
A method has been developed for the determination of arsenic (As) in pharmaceutical and scalp hair samples of kidney failure patients by cloud point extraction (CPE). The scalp hair samples were subjected to microwave-assisted digestion in nitric acid-hydrogen peroxide (2 + 1, v/v). Then, dialysate and digested scalp hair solutions were preconcentrated by CPE using pyronine B as a complexing agent. The resulting complex was entrapped in nonionic surfactant (Triton X-114) prior to its determination by electrothermal atomic absorption spectroscopy. The validity of the CPE method for As was checked by analysis of a certified reference material of human hair and a standard addition method for dialysate solution. The chemical variables affecting the analytical performance of the CPE methods were studied and optimized. After optimization of the complexation and extraction conditions, a preconcentration factor of 52 was obtained for As in 10 mL of dialysate solution and acid digested samples of scalp hair. Under the optimum experimental conditions, the LOD and LOQ of As for the preconcentration of 10 mL of solution were 0.022 and 0.073 microg/L, respectively. The level of As in scalp hair samples of kidney failure patients was higher than in healthy controls.
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Supelco
IGEPAL® CA-630
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Sigma-Aldrich
Triton X-165 solution, Non-ionic, CMC-0.43 mM, suitable for electrophoresis, cloud point >100 °C
Sigma-Aldrich
Triton X-114, laboratory grade
Sigma-Aldrich
Triton X-114
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Pyronin Y, certified by the Biological Stain Commission, Dye content 50 %
Sigma-Aldrich
Pyronin Y, for NA electrophoresis
Sigma-Aldrich
Triton X-405 solution, 70% in H2O
Sigma-Aldrich
Pyronin B, Dye content ≥30 %
Sigma-Aldrich
Pyronin Y, for microscopy (Bot., Fl., Hist.)