Merck
  • Phase inversion of emulsions containing a lipophilic surfactant induced by clay concentration.
Phase inversion of emulsions containing a lipophilic surfactant induced by clay concentration.
Langmuir : the ACS journal of surfaces and colloids (2013-03-01)
Jingchun Zhang, Lu Li, Jun Wang, Jian Xu, Dejun Sun
ABSTRACT
Emulsions stabilized by clay particles and sorbitan monooleate (Span 80) were investigated, and an abnormal phase inversion was observed by increasing the concentration of clay particles in the aqueous phase. At a fixed concentration of Span 80 in the oil phase, the emulsions were oil-in-water (o/w) when the concentration of clay particles in the aqueous phase was low. Surprisingly, the emulsion inverted to water-in-oil (w/o) when the concentration of the hydrophilic clay particles was increased. On the basis of the results of rheological measurements and laser-induced fluorescent confocal microscopy observation, we suggest that this phase inversion is induced by the gel structures formed at high concentration of clay particles. The effects of clay concentration on the stability and the droplet size of these emulsions were also investigated.
Sigma-Aldrich
Paraffin wax, mp ≥65 °C (ASTM D 87)
Supelco
Span® 80, viscosity 1000-2000 mPa.s (20 °C)
Sigma-Aldrich
Paraffin wax, tested according to Ph. Eur., low viscosity
Sigma-Aldrich
Paraffin wax, tested according to Ph. Eur., high viscosity
Sigma-Aldrich
Paraffin wax, pellets, white
Sigma-Aldrich
Paraffin wax, pellets, white, tested according to Ph. Eur.
Sigma-Aldrich
Paraffin wax, meets analytical specification of Ph. Eur., pastilles, white
Sigma-Aldrich
Paraffin oil, puriss., meets analytical specification of Ph. Eur., BP, viscous liquid
Sigma-Aldrich
Paraffin wax, mp 53-58 °C (ASTM D 87)
Sigma-Aldrich
Paraffin wax, mp 58-62 °C (ASTM D 87)
Supelco
Span® 80, suitable for GC
Sigma-Aldrich
Paraffin wax, chunks (large), white
Supelco
Paraffin oil, suitable for IR spectroscopy