Merck
  • Antioxidant mechanism of grape procyanidins in muscle tissues: redox interactions with endogenous ascorbic acid and α-tocopherol.
Antioxidant mechanism of grape procyanidins in muscle tissues: redox interactions with endogenous ascorbic acid and α-tocopherol.
Food chemistry (2013-02-28)
Jacobo Iglesias, Manuel Pazos, Josep Lluís Torres, Isabel Medina
ABSTRACT
The present study investigates the antioxidant mechanism of grape procyanidins and, in particular, their aptitude to establish redox interactions with two important components of the endogenous antioxidant system of muscle tissues, α-tocopherol (α-TOH) and ascorbic acid (AA). To this end, the progress of lipid oxidation was monitored in fish muscle supplemented with grape procyanidins at the concentrations usually employed in antioxidant food applications, and then related to the redox stability of the endogenous α-TOH and AA. In addition to the lipid oxidation protective effect, the incorporation of procyanidins also provided an improvement of the redox stability of the endogenous components in a straight procyanidinic concentration-dependent manner. Results showed the capacity of procyanidins to repair oxidised α-TOH at medium-long term, and to delay the AA depletion. Therefore, such cooperative redox interaction of exogenous procyanidins adequately complements the natural α-TOH regenerative system supplied by AA that is efficient at the early post mortem stages.
Product Description
α-Tocopherol acetate, European Pharmacopoeia (EP) Reference Standard
All-rac-alpha-tocopheryl acetate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
DL-alpha-Tocopherol acetate, analytical standard
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
DL-α-Tocopherol acetate, ≥96% (HPLC)
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Supelco
Tocopheryl Acetate, a, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-α-Tocopherol acetate, tested according to Ph. Eur.