Merck
Imaging of silastic cheek implant penetration into the maxillary sinus.
JAMA otolaryngology-- head & neck surgery (2013-02-23)
Daniel T Ginat, Charles J Schatz
ABSTRACT
Silastic implants are popular for cosmetic cheek augmentation. We describe the computed tomographic (CT) and magnetic resonance imaging (MRI) findings in 5 cases (4 patients) with penetration of the medial aspect of Silastic cheek implants through the anterior maxillary sinus wall. Three cases demonstrated associated sinus mucosal thickening, and 3 cases demonstrated infection surrounding the implant. In 3 cases, the implants migrated into the maxillary sinus, and in 1 case the implant also eroded through the nasoantral wall and extended into the nasal cavity. Maxillary sinus penetration is a potential complication of Silastic cheek implants. The extent of this complication can be characterized by CT and MRI.
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, for oil baths (from −50°C to +200°C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)