Merck
  • Streptococcus sanguinis meningitis following endoscopic ligation for oesophageal variceal haemorrhage.
Streptococcus sanguinis meningitis following endoscopic ligation for oesophageal variceal haemorrhage.
Journal of medical microbiology (2013-02-23)
Yu-Ting Liu, Chin-Fu Lin, Ya-Ling Lee
ABSTRACT
We report a case of acute purulent meningitis caused by Streptococcus sanguinis after endoscopic ligation for oesophageal variceal haemorrhage in a cirrhotic patient without preceding symptoms of meningitis. Initial treatment with flomoxef failed. The patient was cured after 20 days of intravenous penicillin G. This uncommon infection due to S. sanguinis adds to the long list of infectious complications among patients with oesophageal variceal haemorrhage.
Sigma-Aldrich
Penicillin G sodium salt, 96.0-102.0%
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin G sodium salt, ~1650 U/mg
Supelco
Penicillin G potassium salt, VETRANAL®, analytical standard
Sigma-Aldrich
Ceftriaxone disodium salt hemi(heptahydrate), third-generation cephalosporin antibiotic
Sigma-Aldrich
Penicillin G potassium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Penicillin G potassium salt, 95.0 - 102.0%
Ceftriaxone sodium, European Pharmacopoeia (EP) Reference Standard