Merck
  • Colorimetric assay for T4 polynucleotide kinase activity based on the horseradish peroxidase-mimicking DNAzyme combined with λ exonuclease cleavage.
Colorimetric assay for T4 polynucleotide kinase activity based on the horseradish peroxidase-mimicking DNAzyme combined with λ exonuclease cleavage.
Analytica chimica acta (2013-02-23)
Cheng Jiang, Chunyan Yan, Jianhui Jiang, Ruqin Yu
ABSTRACT
T4 polynucleotide kinase (PNK) plays a critical role in various cellular events. Here, we describe a novel colorimetric strategy for estimating the activity of PNK and screening its inhibitors taking advantage of the efficient cleavage of λ exonuclease and the horseradish peroxidase-mimicking DNAzyme (HRPzyme) signal amplification. A label-free hairpin DNA with the sequence of HRPzyme was utilized in the assay. The 5'-hydroxyl terminal of the hairpin DNA was firstly phosphorylated in the presence of PNK and then digested by λ exonuclease. As a result, the blocked 'HRPzyme' sequence of the hairpin DNA was released due to the removal of its completely complementary sequence. Using this strategy, the assay for PNK activity was successfully translated into the detection of HRPzyme. Because of the completely blocking and efficiently releasing of HRPzyme, the colorimetric method exhibited an excellent performance in PNK analysis with a low detection limit of 0.06 U mL(-1) and a wide detection range from 0.06 to 100 U mL(-1). Additionally, the effects of different inhibitors on PNK activity were also evaluated. The proposed strategy holds great potential in the development of high-throughput phosphorylation investigation as well as in the screening of the related drugs.
Product Description
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Polynucleotide Kinase from T4-infected Escherichia coli, 10 units/μL, buffered aqueous glycerol solution