Merck
  • Performance enhancement of ZnO photocatalyst via synergic effect of surface oxygen defect and graphene hybridization.
Performance enhancement of ZnO photocatalyst via synergic effect of surface oxygen defect and graphene hybridization.
Langmuir : the ACS journal of surfaces and colloids (2013-02-20)
Xiaojuan Bai, Li Wang, Ruilong Zong, Yanhui Lv, Yiqing Sun, Yongfa Zhu
ABSTRACT
ZnO1-x/graphene hybrid photocatalyst was prepared via a facile in-situ reduction of graphene oxide and ZnO1-x surface defect oxide. The hybrid photocatlayst showed enhanced photocatalytic activity for the photodegradation of methylene blue. The photocorrosion of ZnO1-x was successfully inhibited by graphene hybridation. ZnO1-x/graphene hybrid photocatalyst with 1.2 wt % graphene showed the optimized photocatalytic activity. The photocatalytic activity of ZnO1-x/graphene-1.2 wt % under visible and UV light was about 4.6 and 1.2 times that of ZnO1-x sample, respectively. The photocurrent intensity of ZnO1-x under visible and UV light irradiation can be enhanced by 2 and 3.5 times by graphene hybridization. The enhancement of photocatalytic activity and photocurrent intensity in ZnO1-x/graphene was attributed to the synergistic effect between graphene and ZnO1-x for high separation efficiency of photoinduced electron-hole pairs mainly resulting from the promotion of HOMO orbit of graphene and the Oi″ defect level of ZnO1-x in ZnO1-x/graphene.
Sigma-Aldrich
Methylene Blue solution, for microscopy, concentrate according to Ehrlich, concentrated, aqueous solution