Merck
  • The protection afforded by the outer layer to procercoids of Schistocephalus solidus during passage through the stomach lumen of their vertebrate host (Gasterosteus aculeatus).
The protection afforded by the outer layer to procercoids of Schistocephalus solidus during passage through the stomach lumen of their vertebrate host (Gasterosteus aculeatus).
Experimental parasitology (2013-02-16)
J Marwaha, K H Jensen, P J Jakobsen
ABSTRACT
Intestinal cestodes with complex life cycles have to pass through the acid stomach lumen of their vertebrate host(s) in order to reach their preferred site of development. The cestode's tegument is the only organ in constant contact with this hostile environment. Procercoids of Schistocephalus solidus (order Diphyllobothriidea) lose their outer layer on passing through the acidic stomach of their second intermediate host (Gasterosteus aculeatus). We wanted to investigate if the outer layer is an adaptation that enables passage through this hostile environment. We used fish bile to force the procercoid larvae to shed their outer layer. This allowed us to compare the survival of the normal procercoids and the transformed ones when exposed to hydrochloric acid. We observed that the presence of the outer layer significantly improved the survival and active period of the procercoid larvae. Thus we conclude that in cestodes which inhabit the digestive tracts of vertebrates, the outer layer is an adaptation which enables them to pass through the acidic stomach environment of their vertebrate host(s).
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, for GC derivatization, LiChropur