Merck
  • Sensitive chemi-sensor for environmental applications as marker of chloroform in aqueous solution.
Sensitive chemi-sensor for environmental applications as marker of chloroform in aqueous solution.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-02-13)
Sher Bahadar Khan, M Faisal, Mohammed M Rahman, Kalsoom Akhtar, Abdullah M Asiri, Khalid A Alamry
ABSTRACT
Sensor technology is a talented progression in the fundamental science and has vital role in the environmental and health monitoring. A lot of sensor materials with different sensitivities have been exposed but the prospective for industrial development as sensors and environmental supplement are still limited. Chloroform chemi-sensor has been fabricated by proficient exploitation of CuO microsphere as redox mediator. CuO microsphere showed good sensitivity (1.497 μM cm(-2) mM(-1)) and lower limit of detection (LOD=0.08 mM) with linear dynamic range (LDR) of 0.13 mM to 0.66 M in short response time (10.0 s). The hydrothermally synthesized CuO microsphere were characterized by using field emission scanning electron microscopy (FE-SEM), X-ray diffraction (XRD), Fourier transform infrared spectroscopy (FTIR) and UV-vis spectra. XRD and FTIR approved that synthesized product is well crystalline, single phase monoclinic CuO. Morphological investigations revealed that CuO composed of nano-block which arranged and structured themselves in a proper manner to give sphere like micro-structures.
Sigma-Aldrich
Copper(II) oxide, 99.999% trace metals basis
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Copper(II) oxide on alumina, 14-30 mesh, extent of labeling: 13 wt. % loading
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Copper(II) oxide, needles, mixture of CuO and Cu2O, ACS reagent
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Copper(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Copper(II) oxide, powder, <10 μm, 98%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer