Merck
  • Multifunctionalization of alkenes via aerobic oxynitration and sp3 C-H oxidation.
Multifunctionalization of alkenes via aerobic oxynitration and sp3 C-H oxidation.
Chemical communications (Cambridge, England) (2013-02-12)
Tsuyoshi Taniguchi, Yuki Sugiura, Takashi Hatta, Atsushi Yajima, Hiroyuki Ishibashi
ABSTRACT
A method for direct functionalization of three positions including an unactivated C-H bond of aliphatic alkenes using tert-butyl nitrite and molecular oxygen to give γ-lactols has been developed. The present reaction proceeds through a sequence of radical processes involving oxynitration followed by aerobic oxidation of an sp(3) C-H bond. This multifunctionalization reaction requires neither metallic reagents nor photolysis and proceeds under mild conditions.
Sigma-Aldrich
Butyl nitrite, 95%