Merck
Bromate adsorption using Fe-pillared bentonite.
Environmental technology (2013-02-12)
Shilong He, Dandan Zhang, Li Gu, Shenghua Zhang, Xin Yu
ABSTRACT
Bromate is an emerging hazardous substance in drinking water. In this study, the removal ofbromate by Fe-pillared bentonite was investigated using various experimental parameters: contact time, initial concentration (Co), temperature, initial pH and competing anions. The adsorption ofbromate followed the pseudo-second-order kinetic better than it followed other kinetic models, and the pseudo-second-order kinetic study showed that equilibrium could be achieved within 60 min. Equilibrium isotherms were analyzed by Freundlich, Langmuir, Redlich-Peterson and Toth isotherm models. The Toth and Redlich-Peterson models better represented the bromate adsorption. Results also indicated that, other than the competing anions and solution pH, temperature was a key parameter affecting adsorption. It was ultimately concluded that Fe-pillared bentonite was effective at removing bromate from water.
Sigma-Aldrich
Bentonite