Merck
  • Entrapped cell system for decentralized hospital wastewater treatment: inhibitory effect of disinfectants.
Entrapped cell system for decentralized hospital wastewater treatment: inhibitory effect of disinfectants.
Environmental technology (2013-02-12)
Sumana Siripattanakul-Ratpukdi
ABSTRACT
This study aims to improve decentralized hospital wastewater treatment inhibited by disinfectants by using calcium alginate cell entrapment technique. The effects of disinfectant types (glutaraldehyde, povidone iodine (PI) and a potassium hydroxide solution) and disinfectant concentrations, cell entrapment conditions (cell-to-matrix ratios) and cell loadings were investigated. The batch experiments were conducted using synthetic wastewater with initial chemical oxygen demand (COD) of approximately 370 mg/L and acclimated activated sludge. Among three disinfectants, PI substantially affected the wastewater treatment efficiency (inhibition of 40%) while other disinfectants exhibited inhibition effects of less than 9%. The results also indicated that the entrapped cells obviously performed better than the free cells. The cell-to-matrix ratio of 1:20 (v/v) provided the highest treatment efficiency of 86% (inhibition of 9%) while the free cell system had inhibition of 47%. The system at the entrapped cell loading of 2000 mg/L performed the highest COD removal of 62% for ten-cycle sequencing batch operation. A scanning electron microscope image provided information on entrapped cell structure subjected to the disinfectant.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Poly(vinylpyrrolidone)–Iodine complex
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets