Merck
  • On the existence of "L-threonine formate", "L-alanine lithium chloride" and "bis L-alanine lithium chloride" crystals.
On the existence of "L-threonine formate", "L-alanine lithium chloride" and "bis L-alanine lithium chloride" crystals.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-02-07)
A M Petrosyan, V V Ghazaryan, M Fleck
ABSTRACT
We argue that the recently reported crystals "L-threonine formate" as well as "L-alanine lithium chloride" and "bis L-alanine lithium chloride" actually are the well-known crystals L-threonine and L-alanine, respectively.
Product Description
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride solution, 8 M, ≥99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, puriss. p.a., anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, Vetec, reagent grade
Sigma-Aldrich
Lithium chloride, ≥99%