Merck
  • Synthesis of porcine brain natriuretic peptide-32 using silver tetrafluoroborate as a new deprotecting reagent of the S-trimethylacetamidomethyl group.
Synthesis of porcine brain natriuretic peptide-32 using silver tetrafluoroborate as a new deprotecting reagent of the S-trimethylacetamidomethyl group.
Chemical & pharmaceutical bulletin (1990-01-01)
M Yoshida, K Akaji, T Tatsumi, S Iinuma, Y Fujiwara, T Kimura, Y Kiso
ABSTRACT
Silver tetrafluoroborate (AgBF4) in trifluoroacetic acid (TFA) has been found to cleave the S-trimethylacetamidomethyl (Tacm) group or the S-acetamidomethyl (Acm) group without affecting other functional groups in the peptide chain. Newly isolated porcine brain natriuretic peptide-32 (pBNP-32) was synthesized using AgBF4 and Cys(Tacm) derivatives. The synthetic pBNP-32 had the highest chick rectum relaxant activity among the ANP-BNP families.
Sigma-Aldrich
Silver tetrafluoroborate, ≥99.99% trace metals basis
Sigma-Aldrich
Silver tetrafluoroborate, 98%