Merck
  • Randomized, controlled, evaluator-blinded studies conducted to compare the efficacy and tolerability of 3 over-the-counter acne regimens in subjects with mild or moderate acne.
Randomized, controlled, evaluator-blinded studies conducted to compare the efficacy and tolerability of 3 over-the-counter acne regimens in subjects with mild or moderate acne.
Journal of drugs in dermatology : JDD (2013-02-05)
Lawrence Green, Leon H Kircik, Jennifer Gwazdauskas
ABSTRACT
Acne vulgaris is a common dermatologic disease that most frequently affects adolescents and young adults. Over-the-counter (OTC) acne treatment regimens are increasingly being used by individuals with acne. To compare the efficacy, user satisfaction, and tolerability of the OTC regimens MaxClarity™, Proactiv®, and Murad® in the treatment of mild and moderate acne. Two randomized, evaluator-blinded, split-face studies were conducted, each involving 20 subjects with acne, to evaluate MaxClarity against Proactiv (study 401) and MaxClarity against Murad (study 404) over 8 weeks. Clinically and statistically significant reductions in acne lesion counts were achieved at 8 weeks compared with baseline for each regimen using MaxClarity, Proactiv, and Murad. Similar reductions in lesion counts and improvements in Investigator's Static Global Assessment grades were observed between MaxClarity and either Proactiv or Murad, in the respective studies. MaxClarity was well tolerated, with no treatment-related adverse events observed in any treatment group and no discontinuations due to adverse events. Overall, most subjects were satisfied with all study treatments. MaxClarity is an effective alternative to either Proactiv or Murad for use in the treatment of mild and moderate acne.
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Luperox® A75, Benzoyl peroxide, 75%, remainder water
Sigma-Aldrich
Luperox® AFR40, Benzoyl peroxide solution, 40 wt. % in dibutyl phthalate
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland