Merck
  • Microfluidic platforms for RNA interference screening of virus-host interactions.
Microfluidic platforms for RNA interference screening of virus-host interactions.
Lab on a chip (2013-01-31)
Benjamin R Schudel, Brooke Harmon, Vinay V Abhyankar, Benjamin W Pruitt, Oscar A Negrete, Anup K Singh
ABSTRACT
RNA interference (RNAi) is a powerful tool for functional genomics with the capacity to comprehensively analyze host-pathogen interactions. High-throughput RNAi screening is used to systematically perturb cellular pathways and discover therapeutic targets, but the method can be tedious and requires extensive capital equipment and expensive reagents. To aid in the development of an inexpensive miniaturized RNAi screening platform, we have developed a two part microfluidic system for patterning and screening gene targets on-chip to examine cellular pathways involved in virus entry and infection. First, a multilayer polydimethylsiloxane (PDMS)-based spotting device was used to array siRNA molecules into 96 microwells targeting markers of endocytosis, along with siRNA controls. By using a PDMS-based spotting device, we remove the need for a microarray printer necessary to perform previously described small scale (e.g. cellular microarrays) and microchip-based RNAi screening, while still minimizing reagent usage tenfold compared to conventional screening. Second, the siRNA spotted array was transferred to a reversibly sealed PDMS-based screening platform containing microchannels designed to enable efficient cell loading and transfection of mammalian cells while preventing cross-contamination between experimental conditions. Validation of the screening platform was examined using Vesicular stomatitis virus and emerging pathogen Rift Valley fever virus, which demonstrated virus entry pathways of clathrin-mediated endocytosis and caveolae-mediated endocytosis, respectively. The techniques here are adaptable to other well-characterized infection pathways with a potential for large scale screening in high containment biosafety laboratories.
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, for oil baths (from −50°C to +200°C)
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)