Merck
Reactions of buffers in cyanogen bromide-induced ligations.
Nucleosides, nucleotides & nucleic acids (2013-01-31)
Heike Vogel, Claudia Gerlach, Clemens Richert
ABSTRACT
Rapid, template-directed ligation reactions between a phosphate-terminated oligonucleotide and an unphosphorylated reaction partner may be induced by cyanogen bromide (BrCN). Frequently, however, the reaction is low yielding, and even a large excess of the condensing agent can fail to induce quantitative conversions. In this study, we used BrCN to induce chemical primer extension reactions. Here, we report that buffers containing hydroxyl groups react with short oligodeoxynucleotides in the presence of BrCN. One stable adduct between HEPBS buffer and cytosine was characterized by mass spectrometry and NMR after HPLC purification, indicating that a side reaction occurred at this nucleobase. Further, a first example of a primer extension reaction between an unmodified oligodeoxynucleotide as primer and dGMP is reported. Together, our results shed light on the potency, as well as the drawbacks of BrCN as a highly reactive condensing reagent for the ligation of unmodified nucleic acids.
Sigma-Aldrich
BES, ≥99.0% (titration)
SAFC
HEPES
SAFC
HEPES sodium salt
Sigma-Aldrich
2′-Deoxyguanosine 5′-monophosphate disodium salt hydrate, ≥98%
Sigma-Aldrich
HEPES sodium salt solution, BioReagent, 1M, suitable for cell culture
Sigma-Aldrich
Cyanogen bromide, reagent grade, 97%
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
HEPES sodium salt, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES sodium salt, Vetec, reagent grade, 96%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Cyanogen bromide, 99.995% trace metals basis
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture