Merck
  • Microwave-assisted improved synthesis of pyrrolo[2,3,4-kl]acridine and dihydropyrrolo[2,3,4-kl]acridine derivatives catalyzed by silica sulfuric acid.
Microwave-assisted improved synthesis of pyrrolo[2,3,4-kl]acridine and dihydropyrrolo[2,3,4-kl]acridine derivatives catalyzed by silica sulfuric acid.
Molecules (Basel, Switzerland) (2013-01-30)
Chengpao Cao, Changliang Xu, Wei Lin, Xuemei Li, Minghua Hu, Juxian Wang, Zhibin Huang, Daqing Shi, Yucheng Wang
ABSTRACT
An improved synthesis of multifunctionalized pyrrolo[2,3,4-kl]acridine derivatives with different substituted patterns using silica sulfuric acid (SSA) as a heterogeneous catalyst under microwave irradiation conditions was developed. The reaction could be conducted by using readily available and inexpensive substrates within short periods of 12-15 min. under microwave irradiation. Compared with the conventional methods, the remarkable advantages of this method are milder reaction conditions, operational simplicity, higher yields, short reaction times, and an environmentally friendly procedure.
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC