Merck
  • In vitro dermal absorption of di(2-ethylhexyl) adipate (DEHA) in a roll-on deodorant using human skin.
In vitro dermal absorption of di(2-ethylhexyl) adipate (DEHA) in a roll-on deodorant using human skin.
Journal of toxicology and environmental health. Part A (2013-01-30)
Simon Ningsun Zhou, Richard P Moody, Bio Aikawa, Anna Yip, Bing Wang, Jiping Zhu
ABSTRACT
In vitro dermal absorption experiments were conducted using a roll-on deodorant that contains 1.56% di(2-ethylhexyl) adipate (DEHA), a plasticizer widely used in consumer products. Human skin specimens were fitted in Bronaugh flow-through Teflon diffusion cells. The diffusion cells were maintained at 32 °C to reflect the skin temperature. Two amounts (low dose: 5 mg of the product; high dose: 100 mg) were applied, in triplicate, each on four different human skins. DEHA was determined in the receiver solution at 6-h intervals, using headspace solid-phase microextraction gas chromatography-mass spectrometry (GC-MS). After 24 h, the experiment was terminated and masses of DEHA in the skin depot, skin wash, and upper and lower chambers of the diffusion cell were determined. A significant portion of applied DEHA, 28% in the low amount application and 34% in the high one, was found in the skin depot. In comparison, only 0.04% and 0.002% of applied DEHA were found in the receiver solutions for the low and high doses, respectively. Under our experimental conditions, an apparent steady-state flux of low DEHA mass penetrating from skin into the receiver solution was observed with a penetration rate of 2.2 ng/cm(2)/h for both the low and high doses. The average mass recovery was 81% for the low dose application and 56% for the high dose.
Product Description
Supelco
Di(2-ethylhexyl)adipate solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Bis(2-ethylhexyl) adipate, ≥97.0% (GC)
Supelco
Bis(2-ethylhexyl) adipate, Selectophore, ≥99.0%
Sigma-Aldrich
Bis(2-ethylhexyl) adipate, ≥99%
Supelco
Bis(2-ethylhexyl) adipate, 99%