Merck
Stabilization of oxidovanadium(IV) by organic radicals.
Dalton transactions (Cambridge, England : 2003) (2013-01-29)
Suman Kundu, Suvendu Maity, Amarendra Nath Maity, Shyue-Chu Ke, Prasanta Ghosh
ABSTRACT
o-Imino-p-R'-benzosemiquinone anion radical (L(R')(IS)(˙-)) complexes of oxidovanadium(IV) of type [(L(1)(R-))(VO(2+))(L(R')(IS)(˙-))] (R = H, R' = H, 1; R = H, R' = -CMe(3), 2; R = -CMe(3), R' = H, 3 and R = -CMe(3), R' = -CMe(3), 4) incorporating the redox-innocent tridentate NNO-donor L(1)(R-) ligands (L(1)(R)H = 2,4-di-R-6-{(2-(pyridin-2-yl)hydrazono)methyl}phenol) were isolated and substantiated by elemental analyses, IR, mass, NMR and UV-vis spectra including the single crystal X-ray structure determinations. The V-O(phenolato) (cis to the V=O) lengths spanning 1.905(3)-1.9355(15) Å in 1-4 are consistent with the coordination to the [VO](2+) state. The V-O(IS) (trans to the V=O) lengths, 2.1505(17)-2.1869(15) Å, in 1-4 are longer due to the trans influence of the V=O bond. The V-N(IS) lengths, 1.906(3)-1.924(2) Å, in 1-4 are comparatively shorter due to the higher affinity of the paramagnetic [VO](2+) ion towards the L(R')(IS)(˙-) anion radicals. Density functional theory (DFT) calculations using B3LYP, B3PW91 and PBE1PBE functionals on 1 and 2 authenticated that the closed shell singlet (CSS) solutions (dianionic o-amido-p-R'-phenolates (L(R')(AP)(2-)) coordinated to VO(3+), Type I) of 1-4 are unstable with respect to the open shell singlet (OSS) perturbations. Broken symmetry, BS (1,1) M(s) = 0 (L(R')(IS)(˙-) coordinated to the VO(2+) ion, Type III) solutions of 1-4 are stable and reproduce the experimental bond parameters. Frozen glasses EPR spectra of [1-4](+) ions (e.g. g(||) = 1.948, g(⊥) = 1.978, A(||) = 184 (22 G), A(⊥) = 62(15 G) for [2](+)) and unrestricted DFT calculations on [1](+), [2](+), [1](-) and [2](-) ions using doublet spin state elucidated that the reversible anodic waves at [0.15-0.31] V of 1-4 complexes are due to the oxidation of L(R')(IS)(˙-) generating [(L(1)(R-))(VO(2+))(L(R')(IQ))]+ complexes (L(R')(IQ) = o-imino-p-R'-benzoquinone) coordinated to the [VO](2+) ion (Type V) while the irreversible cathodic waves at -[1.08-1.49] V are due to the formation of unstable [(L(1)(R-))(VO(2+))(L(R')(AP)(2-))](-) complexes (Type II). The second anodic waves at [0.76-0.89] V are assigned to a [VO](3+)-[VO](2+) couple affording diamagnetic [(L(1)(R-))(VO(3+))(L(R')(IQ))](2+), [1-4](2+) complexes (Type VI) which are identified by UV-vis spectra, DFT and time dependent (TD) DFT calculations. Spectro-electrochemical measurements and TD DFT calculations on 1 and 2 disclosed that lower energy electronic absorption bands of 1-4 are due to the LMCT and CSS-OSS perturbation which disappear in [1-4](+) ions. [1-4](+) absorb at 600-650 nm due to d-d transitions and MLCT which are absent in VO(3+) complexes, [1-4](2+).
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Pyridine hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Pyridine hydrochloride, 98%
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine hydrobromide, 98%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%