Merck
  • The anti-cancer activity of a cationic anti-microbial peptide derived from monomers of polyhydroxyalkanoate.
The anti-cancer activity of a cationic anti-microbial peptide derived from monomers of polyhydroxyalkanoate.
Biomaterials (2013-01-25)
Stephen O'Connor, Emilia Szwej, Jasmina Nikodinovic-Runic, Aisling O'Connor, Annette T Byrne, Marc Devocelle, Norma O'Donovan, William M Gallagher, Ramesh Babu, Shane T Kenny, Manfred Zinn, Qun Ren Zulian, Kevin E O'Connor
ABSTRACT
The biodegradable polymer medium chain length polyhydroxyalkanoate (mclPHA), produced by Pseudomonas putida CA-3, was depolymerised and the predominant monomer (R)-3-hydroxydecanoic acid (R10) purified. R10 was conjugated to a d-peptide DP18 and its derivatives. All peptides conjugated with R10 exhibited greater anti-cancer activity compared to the unconjugated peptides. Unconjugated and conjugated peptides were cytocidal for cancer cells. Conjugation of R10 to peptides was essential for enhanced anti-proliferation activity, as unconjugated mixes did not result in enhancement of anti-cancer activity. The conjugation of R10 resulted in more rapid uptake of peptides into HeLa and MiaPaCa cells compared to unconjugated peptide. Both unconjugated and R10 conjugated peptides localized to the mitochondria of HeLa and MiaPaCa cells and induced apoptosis. Peptide conjugated with a terminally hydroxylated decanoic acid (ω-hydroxydecanoic acid) exhibited 3.3 and 6.3 fold higher IC(50) values compared to R10 conjugated peptide indicating a role for the position of the hydroxyl moiety in enhancement of anti-cancer activity. Conjugation of decanoic acid (C10) to peptides resulted in similar or higher IC(50) values compared to R10 conjugates but C10 conjugates did not exhibit any cancer selectivity. Combination studies showed that R10DP18L exhibited synergy with cisplatin, gemcitabine, and taxotere with IC(50) values in the nanomolar range.
Sigma-Aldrich
Sodium decanoate, ≥98%
Sigma-Aldrich
(±)-3-Hydroxydecanoic acid, ≥98%
Supelco
Decanoic acid, analytical standard
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Decanoic acid, ≥98.0%