Merck
  • [Purification and characteristics of methanol dehydrogenase of Methylobacterium nodulans rhizosphere phytosymbionts].
[Purification and characteristics of methanol dehydrogenase of Methylobacterium nodulans rhizosphere phytosymbionts].
Prikladnaia biokhimiia i mikrobiologiia (2013-01-22)
T A Kuznetsova, A P Beschastnyĭ, O N Ponomareva, Iu A Trotsenko
ABSTRACT
Methanol dehydrogenase (MDG) of the facultative methylotrophic phytosymbiont Methylobacterium nodulans has been purified for the first time to an electrophoretically homogeneous state and characterized. The native protein with a molecular mass of 70 kDa consists of large (60 kDa) and small (6 kDa) subunits. The purified protein displayed a specter identical to that of pyrochinolinchinon (PCC)-containing MDGs (pI 8.7, pH optimum in the range 9-10). The enzyme was inactive in the absence of ammonium or methylamine and exhibited a wide substrate specificity with regard to C1-C2 alcohols with the highest affinity to methanol (K(M) = 70 mM), but it did not oxidize benzyl and secondary alcohols. The apparent values of K(M) to primary alcohols increased with the length of the carbonic chain. The enzyme was characterized by a high stability level even in the absence of a substrate. An immobilized enzyme was used for amperometric methanol detection.
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Methylamine-13C hydrochloride, 99 atom % 13C
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methylamine, ≥99.0%
Sigma-Aldrich
Methylamine-15N hydrochloride, 98 atom % 15N
Sigma-Aldrich
Methylamine hydrochloride, ≥98%