Merck
  • Can boron and nitrogen co-doping improve oxygen reduction reaction activity of carbon nanotubes?
Can boron and nitrogen co-doping improve oxygen reduction reaction activity of carbon nanotubes?
Journal of the American Chemical Society (2013-01-16)
Yu Zhao, Lijun Yang, Sheng Chen, Xizhang Wang, Yanwen Ma, Qiang Wu, Yufei Jiang, Weijin Qian, Zheng Hu
ABSTRACT
Two kinds of boron and nitrogen co-doped carbon nanotubes (CNTs) dominated by bonded or separated B and N are intentionally prepared, which present distinct oxygen reduction reaction (ORR) performances. The experimental and theoretical results indicate that the bonded case cannot, while the separated one can, turn the inert CNTs into ORR electrocatalysts. This progress demonstrates the crucial role of the doping microstructure on ORR performance, which is of significance in exploring the advanced C-based metal-free electrocatalysts.
Sigma-Aldrich
Boron, crystalline, 1 cm, 99.7% trace metals basis
Sigma-Aldrich
Boron, crystalline, −60 mesh, 99% trace metals basis
Sigma-Aldrich
Boron, ≥95% (boron), amorphous powder