Merck
  • Immunomodulatory effects of hexane insoluble fraction of Ficus septica Burm. F. in doxorubicin-treated rats.
Immunomodulatory effects of hexane insoluble fraction of Ficus septica Burm. F. in doxorubicin-treated rats.
Asian Pacific journal of cancer prevention : APJCP (2013-01-16)
Agung Endro Nugroho, Adam Hermawan, Kunti Nastiti, Suven, Pritha Elisa, Tony Hadibarata, Edy Meiyanto
ABSTRACT
The use of chemotherapeutics induces cardiotoxicity and affects immune functions, therefore development of combinatorial agents against cardiotoxicity and immunosuppression needs to be explored. Previous studies of the hexane insoluble fraction (HIF) of an ethanolic extract of Ficus septica leaves showed anticancer effects singly and in combination with doxorubicin on T47D breast cancer cells. In this present study, it was evaluated for its immunomodulatory activities in doxorubicin-treated rats. Thirty male Sprague Dawley rats were divided into five groups consisting of six rats each as follows: Group 1, receiving oral saline 10 ml/kg BW (control group); Group 2, receiving HIF dose 750 mg/kg BW orally, once daily; Group 3, receiving HIF dose 1.500 mg/kg BW orally, once daily; Group 4, given oral saline 10 ml/kg BW (normal group); Group 5, receiving HIF dose 1.500 mg/kg BW orally, once daily. The rats of group 1-3 were intramuscularly administered with doxorubicin at a dose of 4.67 mg/kg BW at the days 1 and 4 to suppress immune functions. Concomitantly, the rats were treated with saline or HIF for seven consecutive days (1 to 7). Treatment of HIF succeeded in reducing side effects of doxorubicin based on increasing lymphocyte density and phagocytosis activity and capacity of macrophages, as well as increasing the CD8+blood level and decreasing spleen IL-10 expression. Hexane insoluble fraction of of ethanolic extract of Ficus septica leaves has potential as a protective agent combined with doxorubicin.
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane-1-13C, 99 atom % 13C
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)