Merck
  • Resistance of brain glucose metabolism to thiopental-induced CNS depression in newborn piglets.
Resistance of brain glucose metabolism to thiopental-induced CNS depression in newborn piglets.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience (2013-01-12)
Bernd Walter, Michael Eiselt, Paul Cumming, Guoming Xiong, Rainer Hinz, Susanne Uthe, Peter Brust, Reinhard Bauer
ABSTRACT
The transition from mild sedation to deep anaesthesia is marked by the phenomenon of burst suppression (BS). FDG-PET studies show that the cerebral metabolic rate for glucose (CMRglc) declines dramatically with onset of BS in the adult brain. Global CMRglc increases substantially in the post-natal period and achieves its maximum in preadolescence. However, the impact of post-natal brain development on the vulnerability of CMRglc to the onset of BS has not been documented. Therefore, cerebral blood flow and metabolism were measured using a variant of the Kety-Schmidt method, in conjunction with quantitative regional estimation of brain glucose uptake by FDG-PET in groups of neonate and juvenile pigs, under a condition of light sedation or after induction of deep anaesthesia with thiopental. Quantification of simultaneous ECoG recordings was used to establish the correlation between anaesthesia-related changes in brain electrical activity and the observed cerebrometabolic changes. In the condition of light sedation the magnitude of CMRglc was approximately 20% higher in the older pigs, with the greatest developmental increase evident in the cerebral cortex and basal ganglia (P<0.05). Onset of BS was associated with 20-40% declines in CMRglc. Subtraction of the mean parametric maps for CMRglc showed the absolute reductions in CMRglc evoked by thiopental anaesthesia to be two-fold greater in the pre-adolescent pigs than in the neonates (P<0.05). Thus, the lesser suppression of brain energy demand of neonate brain during deep anaesthesia represents a reduced part of thiopental suppressing brain metabolism in neonates.
Product Description
Supelco
(±)Thiopental solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
Sigma-Aldrich
(±)-Thiopental