Merck
  • Three dimensional imaging of porosity and tracer concentration distributions in a dolostone sample during diffusion experiments using X-ray micro-CT.
Three dimensional imaging of porosity and tracer concentration distributions in a dolostone sample during diffusion experiments using X-ray micro-CT.
Journal of contaminant hydrology (2013-01-10)
H M D Agbogun, Tom A Al, Esam M A Hussein
ABSTRACT
X-ray micro-computed tomography (micro-CT) techniques for measuring the three-dimensional (3-D) distributions of diffusion-accessible porosity (φ(d)) and temporal tracer-concentrations (C(t)) within a dolostone sample subjected to solute diffusion are developed and tested in this work. The φ(d) and C(t) measurements are based on spatially resolved changes in X-ray attenuation coefficients in sequentially acquired 3-D micro-CT datasets using two (calibration and relative) analytical approaches. The measured changes in X-ray attenuation coefficient values are a function of the mass of X-ray absorbing potassium-iodide tracer present in voxels. Mean φ(d) values of 3.8% and 6.5% were obtained with the calibration and the relative approaches, respectively. The detection limits for φ(d) measurements at individual voxel locations are 20% and 36% with the calibration and the relative methods, respectively. The detection limit for C(t) are 0.12 M and 0.22 M with the calibration and the relative approaches, respectively. Results from the calibration method are affected by a beam-hardening artifact and although results from the relative approach are not affected by the artifact, they are subject to high detection limits. This work presents a quantitative assessment of micro-CT data for studies of solute transport. Despite limitations in precision and accuracy, the method provides quantitative 3-D distributions of φ(d) and C(t) that reflect solute diffusion in heterogeneous porous geologic media.
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Gold etchant, standard