Merck
  • [Sensitivity of Pseudomonas chlororaphis to antibiotics and chemical tools of plant protection].
[Sensitivity of Pseudomonas chlororaphis to antibiotics and chemical tools of plant protection].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2013-01-09)
V V Shepelevich, E A Kiprianova, L V Iaroshenko, L V Avdeeva
ABSTRACT
Examination of sensitivity of 10 Pseudomonas chlororaphis strains belonging to different subspecies to 54 antibiotics has shown that all studied representatives of Pchlororaphis subsp. chlororaphis, P. chlororaphis subsp. aureofaciens and Pchlororaphis subsp. aurantiaca were sensitive to aminoglycoside antibiotics and fluoroquinolones derivatives. Only part of studied strains has shown sensitivity to some beta-lactam antibiotics, imipeneme and meropeneme. In contrast to representatives of two other subspecies both strains of P. chlororaphis subsp. chlororaphis proved to be sensitive to chlortetracycline and cefepime that allows to consider this difference as the characteristic useful for differentiation. All studied P. chlororaphis strains were resistant to chemical fungicides (Scor and Svitch) and the insect growth regulators (Match, Lufox, Engio, Actellik). Resistance of bacteria to these herbicides gives evidence that their combined use is possible.
Product Description
Millipore
Chlortetracycline Selective Supplement, suitable for microbiology
Pricing and availability is not currently available.
Sigma-Aldrich
Chlortetracycline hydrochloride, ≥91.0% dry basis (HPLC)
Pricing and availability is not currently available.
Supelco
Chlortetracycline hydrochloride, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Chlortetracycline hydrochloride, BioReagent, suitable for fluorescence, from Streptomyces aureofaciens, ≥85.0% (HPLC)
Pricing and availability is not currently available.