Merck
  • Temperature- and pH-responsive nanoparticles of biocompatible polyurethanes for doxorubicin delivery.
Temperature- and pH-responsive nanoparticles of biocompatible polyurethanes for doxorubicin delivery.
International journal of pharmaceutics (2012-12-25)
Anning Wang, Hui Gao, Yanfang Sun, Yu-long Sun, Ying-Wei Yang, Guolin Wu, Yinong Wang, Yunge Fan, Jianbiao Ma
ABSTRACT
A series of temperature- and pH-responsive polyurethanes based on hexamethylene diisocyanate (HDI) and 4,4'-diphenylmethane diisocyanate (MDI) were synthesized by a coupling reaction with bis-1,4-(hydroxyethyl) piperazine (HEP), N-methyldiethanolamine (MDEA) and N-butyldiethanolamine (BDEA), respectively. The chemical structure, molecular weight, thermal property and crystallization properties were characterized by Fourier transform infrared (FT-IR) spectroscopy, nuclear magnetic resonance (NMR) spectroscopy, gel permeation chromatography (GPC), differential scanning calorimetry (DSC) and X-ray diffraction (XRD) spectroscopy. The resulting polyurethanes were then used to prepare nanoparticles either by direct dispersion method or dialysis method. Their pH and temperature responsibilities were evaluated by optical transmittance and size measurement in aqueous media. Interestingly, HDI-based and MDI-based polyurethanes exhibited different pH and temperature responsive properties. Nanoparticles based on HDI-HEP and HDI-MDEA were temperature-responsive, while MDI-based biomaterials were not. All of them showed pH-sensitive behavior. The possible responsive mechanism was investigated by (1)H NMR spectroscopy. The cytotoxicity of the polyurethanes was evaluated using methylthiazoletetrazolium (MTT) assay in vitro. It was shown that the HDI-based polyurethanes were non-toxic, and could be applied to doxorubicin (DOX) encapsulation. The experimental results indicated that DOX could be efficiently encapsulated into polyurethane nanoparticles and uptaken by Huh-7 cells. The loaded DOX molecules could be released from the drug-loaded polyurethane nanoparticles upon pH and temperature changes, responsively.
Sigma-Aldrich
Hexamethylene diisocyanate, purum, ≥98.0% (GC)
Sigma-Aldrich
Hexamethylene diisocyanate, puriss., ≥99.0% (GC)
Supelco
4,4′-MDI, analytical standard
Sigma-Aldrich
4,4′-Methylenebis(phenyl isocyanate), 98%