Merck
  • Cucurbit[5]uril-metal complex-induced room-temperature phosphorescence of α-naphthol and β-naphthol.
Cucurbit[5]uril-metal complex-induced room-temperature phosphorescence of α-naphthol and β-naphthol.
Dalton transactions (Cambridge, England : 2003) (2012-12-12)
Zhong-Wei Gao, Xing Feng, Lan Mu, Xin-Long Ni, Li-Li Liang, Sai-Feng Xue, Zhu Tao, Xi Zeng, Bodgan E Chapman, Philip W Kuchel, Leonard F Lindoy, Gang Wei
ABSTRACT
Similar to the larger members of the cucurbituril family, such as cucurbit[8]uril (Q[8]), the smallest member, cucurbit[5]uril (Q[5]), can also induce room-temperature phosphorescence (RTP) of α-naphthol (1) and β-naphthol (2). The relationship between the RTP intensity of 1 and 2 and the concentration of Q[5] or Q[8] suggests that the mechanism underlying the Q[5] complex-induced RTP is different from that of the Q[8]-induced RTP for these luminophores. The crystal structures of 1-Q[5]-KI, 2-Q[5]-KI, 1-Q[5]-TlNO(3), and 2-Q[5]-TlNO(3) systems show that in each case Q[5] and the respective metal ions, K(+) or Tl(+), form infinite ···Q[5]-M(+)-Q[5]-M(+)··· chains that surround the luminophores. Although these tube- or wall-like structures are likely destroyed in solution, the key interaction between the convex-shaped outer walls of Q[5] and the plane of the aromatic naphthols, via π···π stacking and C-H···π interactions, is postulated to be essentially maintained leading to a microenvironment that holds the luminophore and the heavy atom perturber together; such a model is supported by the observed Q[5] complex-induced RTP of the above naphthols. With respect to this, a high Q[5]/luminophore concentration was employed in an endeavour to promote the formation of π···π stacking and C-H···π interactions similar to those observed in the crystal structures of the 1- or 2-Q[5]-K(+) and -Tl(+) systems. In keeping with the proposed model, the RTP of each system is quenched when Q[5] is replaced by the alkyl-substituted Q[5] derivatives, decamethylQ[5] and pentacyclohexanoQ[5]. This is in agreement with the substituent groups on the surface of the metal-bond Q[5] obstructing the naphthol molecule from accessing the convex glycouril backbone of Q[5].
Sigma-Aldrich
2-Naphthol, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
2-Naphthol, 99%
Sigma-Aldrich
2-Naphthol, 98%
Sigma-Aldrich
Gold etchant, standard
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%