Merck
Plasma-induced polymerization for enhancing paper hydrophobicity.
Carbohydrate polymers (2012-12-12)
Zhaoping Song, Jiebin Tang, Junrong Li, Huining Xiao
ABSTRACT
Hydrophobic modification of cellulose fibers was conducted via plasma-induced polymerization in an attempt to graft the hydrophobic polymer chains on paper surface, this increasing the hydrophobicity of paper. Two hydrophobic monomers, butyl acrylate (BA) and 2-ethylhexyl acrylate (2-EHA), were grafted on cellulose fibers, induced by atmospheric cold plasma. Various influencing factors associated with the plasma-induced grafting were investigated. Contact-angle measurement, Fourier Transform infrared spectroscopy (FTIR), X-ray photoelectron spectroscopy (XPS) and scanning electron microscope (SEM) were used to ascertain the occurrence of the grafting and characterized the changes of the cellulose fiber after modification. The results showed that the hydrophobicity of the modified paper sheet was improved significantly after the plasma-induced grafting. The water contact angle on the paper surface reached up to 130°. The morphological differences between modified and unmodified samples were also revealed by SEM observation. The resulting paper is promising as a green-based packaging material.
Product Description
Supelco
2-Ethylhexyl acrylate, analytical standard
Sigma-Aldrich
2-Ethylhexyl acrylate, 98%, contains ≥0.001-≤0.11% monomethyl ether hydroquinone as stabilizer
Supelco
Butyl acrylate, analytical standard
Sigma-Aldrich
Butyl acrylate, ≥99%, contains 10-60 ppm monomethyl ether hydroquinone as inhibitor