Merck
  • Application of mineralogical techniques in the study of human lithiasis.
Application of mineralogical techniques in the study of human lithiasis.
Ultrastructural pathology (2012-12-12)
Jose Aguilar-Ruiz, Miguel Angel Arrabal-Polo, Manuel Sierra, Miguel Arrabal-Martin
ABSTRACT
The authors review the mineralogical methods and techniques of analyzing calculi, stony concretions in the body. They discuss the main types of kidney stones (prostate, testicular, salivary, and bile) and the different diagnostic methods in mineralogy. By applying the techniques of optical microscopy and electron microscopy, they describe the different characteristics of human stones, based on extensive experience as evidenced by their numerous studies.
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Sigma-Aldrich
Sodium oxalate, BioXtra