Merck
  • Engineering E. coli for caffeic acid biosynthesis from renewable sugars.
Engineering E. coli for caffeic acid biosynthesis from renewable sugars.
Applied microbiology and biotechnology (2012-11-28)
Haoran Zhang, Gregory Stephanopoulos
ABSTRACT
Caffeic acid is a valuable aromatic compound that possesses many important pharmacological activities. In structure, caffeic acid belongs to the hydroxycinnamic acid family and can be biosynthesized from the aromatic amino acid tyrosine. In the present paper, the caffeic acid biosynthesis pathway was reconstituted in engineered Escherichia coli to produce caffeic acid from simple biomass sugar glucose and xylose. Different engineering approaches were utilized to optimize the production. Specifically, two parallel biosynthesis routes leading from tyrosine to caffeic acid were studied. The copy number of the intermediate biosynthesis genes was varied to find appropriate gene doses for caffeic acid biosynthesis. Three different media, including a MOPS medium, a synthetic medium, and a rich medium, were also examined to improve the production. The highest specific caffeic acid production achieved was 38 mg/L/OD. Lastly, cultivation of engineered E. coli in a bioreactor resulted in a production of 106 mg/L caffeic acid after 4 days.
Product Description
Supelco
trans-Caffeic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeic acid, ≥99.0% (HPLC)
Supelco
Caffeic acid, ≥99.0% (HPLC), powder, light beige
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)