Merck
  • The long-lasting sensitization of primary afferent nociceptors induced by inflammation involves prostanoid and dopaminergic systems in mice.
The long-lasting sensitization of primary afferent nociceptors induced by inflammation involves prostanoid and dopaminergic systems in mice.
Pharmacology, biochemistry, and behavior (2012-11-28)
Cristiane Flora Villarreal, Mani Indiana Funez, Fernando de Queiroz Cunha, Carlos Amílcar Parada, Sérgio Henrique Ferreira
ABSTRACT
In recent years, evidence that sensitization of primary afferent nociceptors is an important event associated with chronic pain has been accumulating. The present study aimed to evaluate the participation of the prostaglandin and sympathetic components in the long-lasting sensitization of nociceptors induced by acute inflammation in mice. The intraplantar administration of carrageenan (100 μg) enhanced the nociceptive response to a small dose of PGE(2) (9 ng/paw) or dopamine (3 μg/paw) up to 30 days later. This long-lasting sensitization is dependent on dopaminergic and prostanoid systems, since the pre-treatment with chlorpromazine (3 μg/paw) or indomethacin (100 μg/paw), but not local (6 μg/paw) or systemic (6 mg/kg) treatment with morphine, prevented its development. In agreement with this idea, the previous intraplantar administration of hyperalgesic doses of PGE(2) or dopamine also induced long-lasting sensitization, which was fully prevented by pretreatment with EP(4) and D(1) antagonists, respectively. In summary, the present work described in mice a long-lasting sensitization of nociceptors, initiated by an acute inflammatory stimulation and dependent on dopaminergic and prostanoid systems. The present data represent new insights on the mechanisms of peripheral sensitization that could contribute to establish the basis of new therapeutic strategies for acute and chronic inflammatory pain.
Product Description
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard