Merck
  • Histopathological effects of waterborne copper nanoparticles and copper sulphate on the organs of rainbow trout (Oncorhynchus mykiss).
Histopathological effects of waterborne copper nanoparticles and copper sulphate on the organs of rainbow trout (Oncorhynchus mykiss).
Aquatic toxicology (Amsterdam, Netherlands) (2012-11-24)
Genan A Al-Bairuty, Benjamin J Shaw, Richard D Handy, Theodore B Henry
ABSTRACT
It is unclear whether copper nanoparticles are more toxic than traditional forms of dissolved copper. This study aimed to describe the pathologies in gill, gut, liver, kidney, brain and muscle of juvenile rainbow trout, Oncorhynchus mykiss, exposed in triplicate to either a control (no added Cu), 20 or 100 μg l(-1) of either dissolved Cu (as CuSO(4)) or Cu-NPs (mean primary particle size of 87 ± 27 nm) in a semi-static waterborne exposure regime. Fish were sampled at days 0, 4, and 10 for histology. All treatments caused organ injuries, and the kinds of pathologies observed with Cu-NPs were broadly of the same type as CuSO(4) including: hyperplasia, aneurisms, and necrosis in the secondary lamellae of the gills; swelling of goblet cells, necrosis in the mucosa layer and vacuole formation in the gut; hepatitis-like injury and cells with pyknotic nuclei in the liver; damage to the epithelium of some renal tubules and increased Bowman's space in the kidney. In the brain, some mild changes were observed in the nerve cell bodies in the telencephalon, alteration in the thickness of the mesencephalon layers, and enlargement of blood vessel on the ventral surface of the cerebellum. Changes in the proportional area of muscle fibres were observed in skeletal muscle. Overall the data showed that pathology from CuSO(4) and Cu-NPs were of similar types, but there were some material-type effects in the severity or incidence of injuries with Cu-NPs causing more injury in the intestine, liver and brain than the equivalent concentration of CuSO(4) by the end of the experiment, but in the gill and muscle CuSO(4) caused more pathology.
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Copper(II) sulfate, ReagentPlus®, ≥99%
Sigma-Aldrich
Copper(II) sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99-100.5% (based on anhydrous substance)
Sigma-Aldrich
Copper(II) sulfate, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Copper(II) sulfate solution, 4 % (w/v) (prepared from copper (II) sulfate pentahydrate)