Merck
  • Intensive buffering can keep pH above 7.2 for over 4 h during apnea: an experimental porcine study.
Intensive buffering can keep pH above 7.2 for over 4 h during apnea: an experimental porcine study.
Acta anaesthesiologica Scandinavica (2012-11-22)
S Höstman, J Engström, G Hedenstierna, A Larsson
ABSTRACT
Ventilation with low tidal volumes reduces mortality in acute respiratory distress syndrome. A further reduction of tidal volumes might be beneficial, and it is known that apneic oxygenation (no tidal volumes) with arteriovenous CO(2) removal can keep acid-base balance and oxygenation normal for at least 7 h in an acute lung injury model. We hypothesized that adequate buffering might be another approach and tested whether tris-hydroxymethyl aminomethane (THAM) alone could keep pH at a physiological level during apneic oxygenation for 4 h. Six pigs were anesthetized, muscle relaxed, and normoventilated. The lungs were recruited, and apneic oxygenation as well as administration of THAM, 20 mmol/kg/h, was initiated. The experiment ended after 270 min, except one that was studied for 6 h. Two animals died before the end of the experiment. Arterial pH and arterial carbon dioxide tension (PaCO(2) ) changed from 7.5 (7.5, 7.5) to 7.3 (7.2, 7.3) kPa, P < 0.001 at 270 min, and from 4.5 (4.3, 4.7) to 25 (22, 28) kPa, P < 0.001, respectively. Base excess increased from 5 (3, 6) to 54 (51, 57) mM, P < 0.001. Cardiac output and arterial pressure were well maintained. The pig, which was studied for 6 h, had pH 7.27 and PaCO(2) 27 kPa at that time. With intensive buffering using THAM, pH can be kept in a physiologically acceptable range for 4 h during apnea.
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
SAFC
Tromethamine
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline