Merck
  • Comprehensive characterization of Annonaceous acetogenins within a complex extract by HPLC-ESI-LTQ-Orbitrap® using post-column lithium infusion.
Comprehensive characterization of Annonaceous acetogenins within a complex extract by HPLC-ESI-LTQ-Orbitrap® using post-column lithium infusion.
Journal of mass spectrometry : JMS (2012-11-14)
Jessica Le Ven, Isabelle Schmitz-Afonso, Guy Lewin, Olivier Laprévote, Alain Brunelle, David Touboul, Pierre Champy
ABSTRACT
Annonaceous acetogenins (AAGs) are a homogenous class of polyketides proposed as environmental neurotoxins. Previous dereplication studies of AAGs were limited by the use of low-resolution mass spectrometers. Only poor information in terms of structures was provided due to the limited fragmentation of protonated or sodium cationized species. An innovative approach, using reversed-phase high-performance liquid chromatography coupled to a hybrid linear ion trap/orbitrap mass spectrometer (LTQ-Orbitrap®), was therefore performed. Sensitivity was enhanced by post-column infusion of lithium, since AAGs have a high affinity for this cation. High level of structural information was obtained from low-energy-collision-induced dissociation fragmentation experiments of lithium-cationized AAGs ([M + Li](+) ions) as demonstrated with purified standards. The method was then applied to a total ethyl-acetate extract prepared from commercial soursop nectar (Annona muricata L.). The sensitivity, mass accuracy and specific fragmentation patterns proved to be particularly useful for characterization of the AAGs. Typical structural identification procedure and unexpected observations for specific structural types are illustrated, with major and minor compounds.
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%