Merck
  • A highly active and coke-resistant steam reforming catalyst comprising uniform nickel-iron alloy nanoparticles.
A highly active and coke-resistant steam reforming catalyst comprising uniform nickel-iron alloy nanoparticles.
ChemSusChem (2012-11-09)
Mitsuru Koike, Dalin Li, Yoshinao Nakagawa, Keiichi Tomishige
ABSTRACT
Doing fine with Ni-Fe: The calcination and reduction of a hydrotalcite precursor containing Ni and Fe ions gives uniform Ni-Fe alloy nanoparticles mixed with Mg(Ni, Fe, Al)O particles. The uniformity of the Ni-Fe alloy nanoparticles is connected to the catalyst's high activity and resistance to coke formation in toluene and phenol steam reforming reactions.
Sigma-Aldrich
Magnesium hydroxide, reagent grade, 95%
Sigma-Aldrich
Magnesium hydroxide, BioUltra, ≥99.0% (KT)