Merck
Aryl-heteroaryl derivatives as novel wake-promoting agents.
Chemical biology & drug design (2012-11-01)
Brigitte Lesur, Yin G Lin, Val R Marcy, Lisa D Aimone, John Gruner, Edward R Bacon, Sankar Chatterjee
ABSTRACT
In search of a next generation molecule to the novel wake-promoting agent modafinil, a series of aryl-heteroayl-derived wakefulness enhancing agents (in rats) was developed. From this work, compound 16 was separated into its enantiomers to profile them individually.
Product Description
Modafinil for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Modafinil, ≥98% (HPLC)
Modafinil, European Pharmacopoeia (EP) Reference Standard