Merck
  • Facile surface PEGylation via tyrosinase-catalyzed oxidative reaction for the preparation of non-fouling surfaces.
Facile surface PEGylation via tyrosinase-catalyzed oxidative reaction for the preparation of non-fouling surfaces.
Colloids and surfaces. B, Biointerfaces (2012-10-30)
Yunki Lee, Kyung Min Park, Jin Woo Bae, Ki Dong Park
ABSTRACT
The control of biological interactions that occur at material-cell/blood interfaces is of great importance to help maximize in vitro and in vivo performance of biomedical devices. PEGylation has been extensively used as an effective surface modification tool that can alter biological responses on device surfaces. Herein, we report a new surface PEGylation method using a tyrosinase-catalyzed oxidative reaction. Tyramine (TA), an enzymatically active phenolic compound, was chemically conjugated to methoxy poly(ethylene glycol) (mPEG). Surface immobilization of mPEG-TA onto various substrates was accomplished simply and rapidly by adding tyrosinase under mild conditions. It was shown that the water contact angles on all surfaces modified with mPEG-TA were decreased, indicating successful introduction of hydrophilic PEG. In addition, the X-ray photoelectron spectroscopy (XPS) spectra demonstrated the differences in the atomic composition on the TiO(2) surface after treatment with mPEG-TA. Non-fouling surfaces prevent non-specific interactions with proteins and cells; consistently, the PEGylated TiO(2) surface clearly showed a decrease in both levels of bovine serum albumin (BSA) adsorption and NIH3T3 cell attachment. Therefore, the facile surface PEGylation using a tyrosinase-catalyzed oxidative reaction should be useful for designing non-fouling surfaces of biomedical devices.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Sigma-Aldrich
Tyramine, 98%, FG
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350
Sigma-Aldrich
Tyramine, ≥98.0%