Merck
  • A single-blinded randomized clinical trial comparing polymyxin B-trimethoprim and moxifloxacin for treatment of acute conjunctivitis in children.
A single-blinded randomized clinical trial comparing polymyxin B-trimethoprim and moxifloxacin for treatment of acute conjunctivitis in children.
The Journal of pediatrics (2012-10-25)
Lee Williams, Yogangi Malhotra, Barbra Murante, Susan Laverty, Steve Cook, David Topa, Dwight Hardy, Hongyue Wang, Francis Gigliotti
ABSTRACT
To perform a randomized controlled trial comparing moxifloxacin hydrochloride with polymyxin B-trimethoprim for the treatment of acute conjunctivitis. Patients ages 1-18 years old with acute conjunctivitis had cultures performed and were randomized to receive either moxifloxacin hydrochloride or polymyxin B-trimethoprim ophthalmic solution for 7 days. Response to treatment was determined by phone query on day 4-6 and by examination with post-treatment conjunctival culture on day 7-10. One hundred and twenty-four patients were enrolled. Eighty patients (65%) had recognized pathogens (55 Haemophilus influenzae, 22 Streptococcus pneumoniae, 4 Moraxella catarrhalis) isolated from their conjunctiva. One hundred fourteen (56/62 moxifloxacin and 58/62 polymyxin B-trimethoprim) completed the 4-6 day evaluation, with 43/56 (77%) of the moxifloxacin group and 42/58 (72%) of the polymyxin B-trimethoprim group clinically cured according to parents (noninferiority test P = .04). Eighty-nine (39/56 moxifloxacin and 50/58 polymyxin B-trimethoprim) patients completed the 7-10 day evaluation. Clinical cure was observed in 37/39 (95%) of the moxifloxacin and 49/51 (96%) of the polymyxin B-trimethoprim treated groups (noninferiority test P ≤ .01). Clinical cure rates for culture positive and negative conjunctivitis were not different. There was no statistically significant difference in bacteriologic cure rates between the 2 groups. Polymyxin B-trimethoprim continues to be an effective treatment for acute conjunctivitis with a clinical response rate that does not differ from moxifloxacin. Use of polymyxin B-trimethoprim for the treatment of conjunctivitis would result in significant cost savings compared with fluoroquinolones.
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O
Sigma-Aldrich
Polymyxin B sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Polymyxin B sulfate salt
Supelco
Polymyxin B solution, 1 mg/mL in H2O, analytical standard
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Trimethoprim, VETRANAL®, analytical standard
Polymyxin B sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications