Merck
  • Determination of t,t-muconic acid in urine samples using a molecular imprinted polymer combined with simultaneous ethyl chloroformate derivatization and pre-concentration by dispersive liquid-liquid microextraction.
Determination of t,t-muconic acid in urine samples using a molecular imprinted polymer combined with simultaneous ethyl chloroformate derivatization and pre-concentration by dispersive liquid-liquid microextraction.
Analytical and bioanalytical chemistry (2012-10-20)
Mohana Krishna Reddy Mudiam, Abhishek Chauhan, Krishna P Singh, Shailendra K Gupta, Rajeev Jain, Ratnasekhar Ch, R C Murthy
ABSTRACT
The present communication describes the preparation and evaluation of a molecularly imprinted polymer (MIP) as a solid-phase extraction (SPE) sorbent and simultaneous ethyl chloroformate (ECF) derivatization and pre-concentration by dispersive liquid-liquid microextraction (DLLME) for the analysis of t,t-muconic acid (t,t-MA) in urine samples using gas chromatography-mass spectrometry. The imprinting polymer was prepared using methacrylic acid as a functional monomer, ethylene glycol dimethacrylate as a cross-linker, 2,2-azobisisobutyronitrile as the initiator and t,t-MA as a template molecule. The imprinted polymer was evaluated for its use as a SPE sorbent by comparing both imprinted and non-imprinted polymers in terms of the recovery of t,t-MA from urine samples. Molecular modelling studies were performed in order to estimate the binding energy and efficiency of the MIP complex formed between the monomer and the t,t-MA. Various factors that can affect the extraction efficiency of MIP, such as the loading, washing and eluting conditions, were optimized; other factors that can affect the derivatization and DLLME pre-concentration were also optimized. MIP in combination with ECF derivatization and DLLME pre-concentration for t,t-MA exhibits good linearity, ranging from 0.125 to 2 μg mL(-1) (R(2) = 0.9971), with limit of detection of 0.037 μg mL(-1) and limit of quantification of 0.109 μg mL(-1). Intra- and inter-day precision was found to be <6%. The proposed method has been proven to be effective and sensitive for the selective pre-concentration and determination of t,t-MA in urine samples of cigarette smokers.
Sigma-Aldrich
trans,trans-Muconic acid, 95%
Sigma-Aldrich
Ethyl chloroformate, purum, ≥98.0% (GC)
Sigma-Aldrich
Sorbic acid, 99.0-101.0% anhydrous basis
Sigma-Aldrich
Ethyl chloroformate, 97%
Sorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sorbic acid, tested according to Ph. Eur.