Merck
  • Biochemical and biophysical response to calcium chloride stress in Aspergillus niger and its role in malachite green degradation.
Biochemical and biophysical response to calcium chloride stress in Aspergillus niger and its role in malachite green degradation.
Cell biochemistry and biophysics (2012-10-19)
Ola M Gomaa, Nabila S Selim, John E Linz
ABSTRACT
Filamentous fungi show great promise in remediation of environmental contaminants such as industrial dyes. In the current study, Aspergillus niger (Genbank ID: JF437542) decolorized 82 % of the test dye malachite green (MG; 50 mg/l) during cultivation for 24 h. The organism decolorized only 6 % of the MG at higher concentration (250 mg MG/l) during the same time period and growth was inhibited at this higher MG concentration. Exposing A. niger to different types of stress resulted in variable impacts on ability to decolorize MG. CaCl2 had the largest positive impact on decolorization. A. niger cultures treated with CaCl2 (1 M) decolorized 46 % of the MG (250 mg/l) in 1 h compared to 6 % in untreated control cultures. CaCl2 also increased catalase production in A. niger which strongly supported a direct relationship between stress response and decolorizing ability. Spectrophotometric measurement confirmed MG decolorization while Fourier transform infrared spectroscopy suggested that biodegradation of MG occurred. Cultures treated with CaCl2 accumulated fewer toxic MG by-products than untreated cultures. CaCl2-induced stress increased the permeability and conductivity of the fungal cell membrane. An observed increase in medium [H(+)] also suggested a change in Ca(2+)/H(+) exchange capacity in the fungal cell. Calcium ions had a pronounced effect on membrane properties and this may have had an important impact on signal transduction. We conclude that A. niger decolorizes MG and that CaCl2 enhances this process; the CaCl2 effect appears to be associated with stress response.
Sigma-Aldrich
Malachite Green oxalate salt, Technical grade
Supelco
Malachite Green oxalate salt, VETRANAL®, analytical standard
Sigma-Aldrich
Malachite Green oxalate salt, for microscopy, crystalline, S. No.: 754
Sigma-Aldrich
Malachite Green oxalate salt, certified by the Biological Stain Commission