Merck
  • Effect of dietary iodine on thyroid hormones and energy blood metabolites in lactating goats.
Effect of dietary iodine on thyroid hormones and energy blood metabolites in lactating goats.
Animal : an international journal of animal bioscience (2012-10-04)
A Nudda, G Battacone, G Bomboi, B Floris, M Decandia, G Pulina
ABSTRACT
Aim of this work was to evaluate if long-term dietary supplementation of potassium iodide (KI) to dairy goats can influence metabolic and hormonal parameters. Thirty Sarda crossbred dairy goats were divided into three groups, which were orally administered 0 (control group; CON), 0.45 (low iodine group; LI) or 0.90 (high iodine group; HI) mg of KI/day, respectively. The daily dose of KI (76.5% of iodine) was administered as salt dissolved in water for 8 weeks. Plasma contents of nonesterified fatty acids (NEFA), urea, glucose, insulin, free triiodothyronine (FT(3)) and thyroxine (FT(4)) were determined weekly. Iodine supplementation increased significantly the FT(3) hormone (P = 0.007) and FT(3)/FT(4) ratio (P = 0.001) and tended to influence the FT(4) hormone (P = 0.059). An iodine level × week of sampling interaction for NEFA (P = 0.013) evidenced a temporary concentration increase in supplemented groups. The 'Revised Quantitative Insulin Sensitivity Check Index' increased with KI supplementation (P ≤ 0.01). Blood urea nitrogen (BUN) and insulin were lowered (P ≤ 0.01) by iodine supplementation (groups LI and HI; P ≤ 0.01). The glucose concentration evidenced an iodine level × week of sampling interaction (P = 0.025) due to an unexpected and temporary increase of its concentration in the CON group. Glucose concentration was decreased by KI supplementation only in LI group (P < 0.05). In conclusion, the daily supplementation of low doses of KI can improve insulin sensitivity and decrease BUN in dairy goats.
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Gold etchant, standard