Merck
  • Chemical composition, olfactory analysis and antibacterial activity of Thymus vulgaris chemotypes geraniol, 4-thujanol/terpinen-4-ol, thymol and linalool cultivated in southern France.
Chemical composition, olfactory analysis and antibacterial activity of Thymus vulgaris chemotypes geraniol, 4-thujanol/terpinen-4-ol, thymol and linalool cultivated in southern France.
Natural product communications (2012-09-18)
Erich Schmidt, Jürgen Wanner, Martina Hiiferl, Leopold Jirovetz, Gerhard Buchbauer, Velizar Gochev, Tania Girova, Albena Stoyanova, Margit Geissler
ABSTRACT
The essential oils of four chemotypes of Thymus vulgaris L. (Lamiaceae) were analyzed for their composition and antibacterial activity to assess their different properties. GC-MS and GC-FID analyses revealed that the essentials oils can be classified into the chemotypes thymol (41.0% thymol), geraniol (26.4% geraniol), linalool (72.5% linalool) and 4-thujanol/terpinen-4-ol (42.2% cis- and 7.3% trans-sabinene hydrate, 6.5 % terpinen-4-ol). The olfactory examination confirmed the explicit differences between these chemotypes. Furthermore, antibacterial activity was investigated against several strains of two Gram-positive (Brochothrix thermosphacta and Staphylococcus aureus) and four Gram-negative food-borne bacteria (Escherichia coli, Salmonella abony, Pseudomonas aeruginosa and P. fragi). All essential oil samples were demonstrated to be highly effective against Gram-positive strains, whereas the impact on Gram-negative microorganisms was significantly smaller, but still considerable. The results obtained indicate that, despite their different properties, the essential oils of selected T. vulgaris chemotypes are potent antimicrobials to be employed as useful additives in food products as well as for therapeutic applications.
Product Description
Supelco
(−)-Linalool, analytical standard
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, ≥97%, FCC, FG
Sigma-Aldrich
L-Linalool, natural, ≥95%, FG
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%
Supelco
Linalool, analytical standard
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
Geraniol, natural, ≥97%, FG
Sigma-Aldrich
Linalool, 97%
Supelco
Geraniol, analytical standard