Merck
  • Microwave-assisted one-step extraction-derivatization for rapid analysis of fatty acids profile in herbal medicine by gas chromatography-mass spectrometry.
Microwave-assisted one-step extraction-derivatization for rapid analysis of fatty acids profile in herbal medicine by gas chromatography-mass spectrometry.
The Analyst (2012-09-13)
Rui-Lin Liu, Jing Zhang, Zhao-Li Mou, Shuang-Li Hao, Zhi-Qi Zhang
ABSTRACT
A rapid and practical microwave-assisted one-step extraction-derivatization (MAED) method was developed for gas chromatography-mass spectrometry analysis of fatty acids profile in herbal medicine. Several critical experimental parameters for MAED, including reaction temperature, microwave power and the amount of derivatization reagent (methanol), were optimized with response surface methodology. The results showed that the chromatographic peak areas of total fatty acids and total unsaturated fatty acids content obtained with MAED were markedly higher than those obtained by the conventional Soxhlet or microwave extraction and then derivatization method. The investigation of kinetics and thermodynamics of the derivatization reaction revealed that microwave assistance could reduce activation energy and increase the Arrhenius pre-exponential factor. The MAED method simplified the sample preparation procedure, shortened the reaction time, but improved the extraction and derivatization efficiency of lipids and reduced ingredient losses, especially for the oxidization and isomerization of unsaturated fatty acids. The simplicity, speed and practicality of this method indicates great potential for high throughput analysis of fatty acids in natural medicinal samples.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets