Merck
  • Role of plant growth regulators and a saprobic fungus in enhancement of metal phytoextraction potential and stress alleviation in pearl millet.
Role of plant growth regulators and a saprobic fungus in enhancement of metal phytoextraction potential and stress alleviation in pearl millet.
Journal of hazardous materials (2012-09-11)
Firdaus-e-Bareen, Muhammad Shafiq, Sidra Jamil
ABSTRACT
"Assisted phytoextraction" involving application of chemical additives such as plant growth regulators (PGRs) has become a trend in phytoremediation technology. This study identifies a cost-effective, naturally available crude PGR (PGR1) that produces the same effects as the commercial PGR (PGR2), increasing metal uptake by plants and the reduction of metal stress. Assisted phytoextraction by pearl millet (Pennisetum glaucum) from a multi-metal (Cd, Cr, Cu, Fe, Na and Zn) contaminated soil medium with tannery solid waste (TSW) soil amendments of 5 and 10%, was evaluated in a full-factorial pot trial with PGR1, PGR2 and Trichoderma pseudokoningii as factors. The effects of these phytoextraction assistants were measured through dry biomass production, heavy metal uptake, stress tolerance enzymes catalase (CAT) and superoxide dismutase (SOD), soluble protein content of plant, and phytoextraction efficiency. Dry biomass and multi-metal accumulation were the highest in the soil treatments with a combined application of PGR1, PGR2 and T. pseudokoningii and the lowest in the control. The soluble protein contents and the SOD and CAT values were the highest in the 10% TSW treatment provided with PGR2+T. pseudokoningii, while the lowest were in the control. Thus, application of crude PGR in combination with other phytoextraction assistants can increase biomass production as well as multi-metal accumulation in plants. However, the biochemical properties of the plant depend on the level of TSW stress in the soil treatment as well as the type of phytoextraction assistants.
Sigma-Aldrich
6-Benzylaminopurine, suitable for plant cell culture
Sigma-Aldrich
Kinetin, suitable for plant cell culture, crystalline
Sigma-Aldrich
Kinetin, suitable for plant cell culture, BioReagent, amorphous powder
Sigma-Aldrich
6-Benzylaminopurine, ReagentPlus®, ≥99.0% (HPLC)
Sigma-Aldrich
Kinetin, ≥99.0% (HPLC)
Supelco
6-Benzylaminopurine, PESTANAL®, analytical standard
Sigma-Aldrich
Kinetin solution, 1 mg/mL, BioReagent, suitable for plant cell culture