Merck
  • Occurrence of aflatoxin M(1) in some samples of UHT, raw & pasteurized milk from Indian states of Karnataka and Tamilnadu.
Occurrence of aflatoxin M(1) in some samples of UHT, raw & pasteurized milk from Indian states of Karnataka and Tamilnadu.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-09-04)
Vinutha Siddappa, Divyashree Kallenahalli Nanjegowda, Prema Viswanath
ABSTRACT
Aflatoxin M(1) (AFM(1)) is a toxic metabolite found in the milk of lactating animals which have consumed feedstuffs contaminated with aflatoxin B(1). Ultra high temperature treated (UHT) milk is a product which is becoming popular in developing countries like India as there is a lack of proper cold storage or refrigeration facilities. In this study, 45 samples of UHT milk of popular brands prevalent in the market were analyzed for the presence of AFM(1) by reversed phase HPLC using fluorescent detector after cleanup of sample with immunoaffinity columns. All samples of plain UHT milk were positive for AFM(1) and 38% of these contained levels more than 0.5 μg/kg, the maximum permitted limit prescribed by the Codex Alimentarius Commission and by the mandatory regulations of the country, the FSSAI Regulations, 2011. In 62.5% of flavored UHT milk, AFM(1) was below detectable levels (0.02 μgL(-1)). However, 12.5% of these samples also contained levels exceeding the maximum permitted limits. AFM(1) was present in 61.6% of the 52 raw milk samples analyzed from the two states of Karnataka and Tamilnadu with a range of 0.1-3.8 μgL(-1). 17.3% of these samples also exceeded the regulatory limits of the country.
Sigma-Aldrich
Aflatoxin M1, from Aspergillus flavus
Supelco
Aflatoxin M1 solution, 0.5 μg/mL in acetonitrile, analytical standard