Merck
  • Formation of pH-sensitive cationic liposomes from a binary mixture of monoalkylated primary amine and cholesterol.
Formation of pH-sensitive cationic liposomes from a binary mixture of monoalkylated primary amine and cholesterol.
Langmuir : the ACS journal of surfaces and colloids (2012-08-31)
Zhong-Kai Cui, Anne Bouisse, Nicolas Cottenye, Michel Lafleur
ABSTRACT
It has been shown that mixtures of monoalkylated amphiphiles and sterols can form liquid-ordered (lo) lamellar phases. These bilayers can be extruded using conventional methods to obtain large unilamellar vesicles (LUVs) that have very low permeability and a specific response to a given stimulus. For example, pH variations can trigger the release from LUVs formed with palmitic acid and sterols. In the present work, the possibility to form non phospholipid liposomes with mixtures of stearylamine (SA) and cholesterol (Chol) was investigated. The phase behavior of these mixtures was characterized by differential scanning calorimetry, infrared, and (2)H NMR spectroscopy. It is found that this particular mixture can form a lo lamellar phase that is pH-sensitive as the system undergoes a transition from a lo phase to a solid state when pH is increased from 5.5 to 12. LUVs have been successfully extruded from equimolar SA/Chol mixtures. Release experiments as a function of time revealed the relatively low permeability of these systems. The fact that the stability of these liposomes is pH dependent implies that these LUVs display an interesting potential as new cationic carriers for pH-triggered release. This is the first report of non phospholipid liposomes with high sterol content combining an overall positive charge and pH-sensitivity.
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Sigma-Aldrich
Octadecylamine, technical grade, 90%
Sigma-Aldrich
Octadecylamine, ≥99.0% (GC)