Merck
  • A synthetic method to access symmetric and non-symmetric 2-(N,N'-disubstituted)guanidinebenzothiazoles.
A synthetic method to access symmetric and non-symmetric 2-(N,N'-disubstituted)guanidinebenzothiazoles.
Molecules (Basel, Switzerland) (2012-08-28)
Alejandro Cruz, Itzia I Padilla-Martínez, Efrén V García-Báez
ABSTRACT
Symmetric and non-symmetric 2-(N-H, N-methyl, N-ethylenyl and N-aryl)guanidinebenzothiazoles were synthesized from the reaction of ammonia, methylamine, pyrrolidine and aniline with dimethyl benzo[d]thiazol-2-yl-carbonodithioimidate as intermediate. The products were characterized by ¹H-, ¹³C-NMR spectroscopy and three of them by X-ray diffraction analysis. HN-phenyl protons formed intramolecular hydrogen bonds that assist the stereochemistry of the second substituent, whereas the HN-alkyl protons were involved in intermolecular hydrogen bonding.
Sigma-Aldrich
Pyrrolidine, FG
Sigma-Aldrich
Pyrrolidine, ≥99.0%
Sigma-Aldrich
Methylamine-13C hydrochloride, 99 atom % 13C
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
Pyrrolidine, 99%
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Methylamine-15N hydrochloride, 98 atom % 15N
Sigma-Aldrich
Methylamine, ≥99.0%
Sigma-Aldrich
Methylamine hydrochloride, ≥98%